3-Methyl-4-nitrobenzotrifluoride 97%
Dodavatel: Apollo Scientific
PC520502-25G
PC520502-5G
APOSPC520502-25GEA
27180
CZK
APOSPC520502-25G
APOSPC520502-5G
3-Methyl-4-nitrobenzotrifluoride 97%
3-Methyl-4-nitrobenzotrifluoride
Vzorec:
C₈H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00042323 CAS číslo: 67192-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...