Methyl trifluoropyruvate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate
PC5185E-25G
PC5185E-5G
APOSPC5185E-25GEA
953
CZK
APOSPC5185E-25G
APOSPC5185E-5G
Methyl trifluoropyruvate 95%
Methyl trifluoropyruvate
Vzorec:
C₄H₃F₃O₃ MW: 156,06 g/mol Bod varu: 84…86 °C Hustota: 1,529 g/cm³ (25 °C) Bod vzplanutí: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00114936 CAS číslo: 13089-11-7 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...