Tisk…

4-Bromo-2-fluoro-6-nitroaniline

Dodavatel: Apollo Scientific
Varování

PC5180-1G PC5180-25G PC5180-5G
APOSPC5180-1GEA 622 CZK
APOSPC5180-1G APOSPC5180-25G APOSPC5180-5G
4-Bromo-2-fluoro-6-nitroaniline
4-Bromo-2-fluoro-6-nitroaniline
Vzorec: C₆H₄BrFN₂O₂
MW: 235,0100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094271
CAS číslo: 517920-70-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR