Tisk…

N-Methyl-4-(trifluoromethoxy)aniline 95%

Dodavatel: Apollo Scientific
Varování

PC5171G-1G PC5171G-25G PC5171G-5G
APOSPC5171G-1GEA 827 CZK
APOSPC5171G-1G APOSPC5171G-25G APOSPC5171G-5G
N-Methyl-4-(trifluoromethoxy)aniline 95%
N-Methyl-4-(trifluoromethoxy)aniline
Vzorec: C₈H₈F₃NO
MW: 191,15 g/mol
Bod varu: 96…98 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052332
CAS číslo: 41419-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR