Methyl-4,4,4-trifluorocrotonate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC5167-5G
PC5167-10G
PC5167-1G
APOSPC5167-5GEA
5290
CZK
APOSPC5167-5G
APOSPC5167-10G
APOSPC5167-1G
Methyl-4,4,4-trifluorocrotonate 95%
Methyl-4,4,4-trifluorocrotonate
Vzorec:
C₅H₅F₃O₂ Bod varu: 94…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077567 CAS číslo: 85694-31-1 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...