Tisk…

2-Amino-5-fluoro-3-nitrobenzotrifluoride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-2-nitro-6-(trifluoromethyl)aniline

PC5166-5G PC5166-1G PC5166-25G
APOSPC5166-5GEA 2360 CZK
APOSPC5166-5G APOSPC5166-1G APOSPC5166-25G
2-Amino-5-fluoro-3-nitrobenzotrifluoride 97%
2-Amino-5-fluoro-3-nitrobenzotrifluoride
Vzorec: C₇H₄F₄N₂O₂
MW: 224,1125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094266
CAS číslo: 344-29-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR