Tisk…

tert-Butyl (3S,4R)-3-fluoro-4-hydroxypyrrolidine-1-carboxylate 96%

Dodavatel: Apollo Scientific

PC51395-100MG PC51395-250MG
APOSPC51395-100MGEA 12870 CZK
APOSPC51395-100MG APOSPC51395-250MG
tert-Butyl (3S,4R)-3-fluoro-4-hydroxypyrrolidine-1-carboxylate 96%
tert-Butyl (3S,4R)-3-fluoro-4-hydroxypyrrolidine-1-carboxylate
Vzorec: C₉H₁₆FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD23106144
CAS číslo: 1174020-48-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR