Methyl perfluorododecanoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC5136-25G
PC5136-5G
APOSPC5136-25GEA
4070
CZK
APOSPC5136-25G
APOSPC5136-5G
Methyl perfluorododecanoate 95%
Methyl perfluorododecanoate
Vzorec:
C₁₃H₃F₂₃O₂ Bod tání: 54…56 °C Hustota: 1,66 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236616 CAS číslo: 56554-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...