Methyl perfluorodecanoate 94%
Dodavatel: Apollo Scientific
|
Varování
|
PC5135-25G
PC5135-5G
APOSPC5135-25GEA
4630
CZK
APOSPC5135-25G
APOSPC5135-5G
Methyl perfluorodecanoate 94%
Methyl perfluorodecanoate
Vzorec:
C₁₁H₃F₁₉O₂ Bod varu: 193 °C Hustota: 1,497 g/cm³ (25 °C) Bod vzplanutí: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153211 CAS číslo: 307-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...