Tisk…

Methyl pentafluoropropionate 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 2,2,3,3,3-pentafluoropropanoate

PC5130-100G
APOSPC5130-100GEA 4240 CZK
APOSPC5130-100G
Methyl pentafluoropropionate 99%
Methyl pentafluoropropionate
Vzorec: C₄H₃F₅O₂
MW: 178,06 g/mol
Bod varu: 59…60 °C
Hustota: 1,393 g/cm³ (25 °C)
Bod vzplanutí: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000430
CAS číslo: 378-75-6
EINECS: 206-828-0
UN: 3272
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR