Tisk…

Methyl pentafluoromethacrylate 96%

Dodavatel: Apollo Scientific
Varování

PC5128-5G PC5128-1G PC5128-250MG
APOSPC5128-5GEA 23780 CZK
APOSPC5128-5G APOSPC5128-1G APOSPC5128-250MG
Methyl pentafluoromethacrylate 96%
Methyl pentafluoromethacrylate
Vzorec: C₅H₃F₅O₂
MW: 190.07 g/mol
Bod varu: 94…95 °C
Hustota: 1.373 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00153210
CAS číslo: 685-09-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR