(Trifluoromethyl)cyclohexane
Dodavatel: Apollo Scientific
|
Varování
|
PC51228-1G
PC51228-5G
APOSPC51228-1GEA
2380
CZK
APOSPC51228-1G
APOSPC51228-5G
(Trifluoromethyl)cyclohexane
(Trifluoromethyl)cyclohexane
Vzorec:
C₇H₁₁F₃ Bod varu: 104…107 °C Hustota: 1,149 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03412289 CAS číslo: 401-75-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...