Tisk…

4-Bromo-2-(difluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific
Varování

PC51201-100MG PC51201-1G PC51201-500MG PC51201-250MG
APOSPC51201-100MGEA 913 CZK
APOSPC51201-100MG APOSPC51201-1G APOSPC51201-500MG APOSPC51201-250MG
4-Bromo-2-(difluoromethyl)pyridine 97%
4-Bromo-2-(difluoromethyl)pyridine
Vzorec: C₆H₄BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD18257229
CAS číslo: 1211580-54-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR