Tisk…

2-Fluoro-5-formyl-3-picoline

Dodavatel: Apollo Scientific
Varování

PC51112-1G PC51112-5G
APOSPC51112-1GEA 2650 CZK
APOSPC51112-1G APOSPC51112-5G
2-Fluoro-5-formyl-3-picoline
2-Fluoro-5-formyl-3-picoline
Vzorec: C₇H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD06659485
CAS číslo: 884495-04-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR