Methyl heptafluorobutyrate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2,2,3,3,4,4,4-heptafluorobutanoate, Methyl perfluorobutyrate
PC5110-50G
PC5110-5G
APOSPC5110-50GEA
4370
CZK
APOSPC5110-50G
APOSPC5110-5G
Methyl heptafluorobutyrate 98%
Methyl heptafluorobutyrate
Vzorec:
C₅H₃F₇O₂ MW: 228,07 g/mol Bod varu: 80…81 °C Bod tání: –86 °C Hustota: 1,472 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000433 CAS číslo: 356-24-1 EINECS: 206-600-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...