Tisk…

5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid, 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

PC5109-1G PC5109-5G
APOSPC5109-1GEA 1370 CZK
APOSPC5109-1G APOSPC5109-5G
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 97%
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Vzorec: C₉H₁₀BFO₄
MW: 211,98 g/mol
Bod tání: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD08235094
CAS číslo: 874219-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR