4,4-Difluorocyclohexanol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC51054-1G
PC51054-5G
APOSPC51054-1GEA
2070
CZK
APOSPC51054-1G
APOSPC51054-5G
4,4-Difluorocyclohexanol 95%
4,4-Difluorocyclohexanol
Vzorec:
C₆H₁₀F₂O MW: 136,1398 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10000566 CAS číslo: 22419-35-8 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...