6-Bromo-5-fluoro-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
PC51011-1G
PC51011-250MG
PC51011-500MG
PC51011-5G
PC51011-10G
APOSPC51011-1GEA
3540
CZK
APOSPC51011-1G
APOSPC51011-250MG
APOSPC51011-500MG
APOSPC51011-5G
APOSPC51011-10G
6-Bromo-5-fluoro-1H-indazole
6-Bromo-5-fluoro-1H-indazole
Vzorec:
C₇H₄BrFN₂ MW: 215.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18089816 CAS číslo: 1286734-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...