Tisk…

Methyl-4-bromo-2-fluorophenylacetate

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-(4-bromo-2-fluorophenyl)acetate

PC510106-5G
APOSPC510106-5GEA 1580 CZK
APOSPC510106-5G
Methyl-4-bromo-2-fluorophenylacetate
Methyl-4-bromo-2-fluorophenylacetate
Vzorec: C₉H₈BrFO₂
MW: 247,063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483545
CAS číslo: 193290-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR