2,2,2,3',5'-Pentafluoroacetophenone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(3,5-Difluorophenyl)-2,2,2-trifluoroethanone
PC5101-25G
PC5101-5G
PC5101-100G
APOSPC5101-25GEA
2070
CZK
APOSPC5101-25G
APOSPC5101-5G
APOSPC5101-100G
2,2,2,3',5'-Pentafluoroacetophenone
2,2,2,3',5'-Pentafluoroacetophenone
Vzorec:
C₈H₃F₅O MW: 210,1000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01319993 CAS číslo: 845823-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...