Tisk…

6-(3,5-Difluorophenyl)-3-pyridinecarbaldehyde

Dodavatel: Apollo Scientific

PC510062-1G PC510062-5G
APOSPC510062-1GEA 16830 CZK
APOSPC510062-1G APOSPC510062-5G
6-(3,5-Difluorophenyl)-3-pyridinecarbaldehyde
6-(3,5-Difluorophenyl)-3-pyridinecarbaldehyde
Vzorec: C₁₂H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115470
CAS číslo: 898404-56-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR