Tisk…

2,4-Dichloro-8-fluoroquinazoline

Dodavatel: Apollo Scientific

PC510014-1G PC510014-250MG PC510014-5G
APOSPC510014-1GEA 6750 CZK
APOSPC510014-1G APOSPC510014-250MG APOSPC510014-5G
2,4-Dichloro-8-fluoroquinazoline
2,4-Dichloro-8-fluoroquinazoline
Vzorec: C₈H₃Cl₂FN₂
MW: 217,300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954886
CAS číslo: 959237-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR