Tisk…

1-Acetyl-5-fluoro-1H-indazole

Dodavatel: Apollo Scientific

PC510013-5G PC510013-1G
APOSPC510013-5GEA 20300 CZK
APOSPC510013-5G APOSPC510013-1G
1-Acetyl-5-fluoro-1H-indazole
1-Acetyl-5-fluoro-1H-indazole
Vzorec: C₉H₇FN₂O
Storage Temperature: Ambient
MDL Number: MFCD12913626
CAS číslo: 141071-11-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR