Tisk…

2-Bromo-3-fluoroaniline 98%

Dodavatel: Apollo Scientific
Varování

PC5082-5G PC5082-1G PC5082-25G
APOSPC5082-5GEA 705 CZK
APOSPC5082-5G APOSPC5082-1G APOSPC5082-25G
2-Bromo-3-fluoroaniline 98%
2-Bromo-3-fluoroaniline
Vzorec: C₆H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD07369915
CAS číslo: 111721-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR