Tisk…

Methyl-3,5-bis(trifluoromethyl)benzoate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3,5-bis(trifluoromethyl)benzoate

PC5076-25G PC5076-5G PC5076-100G
APOSPC5076-25GEA 2070 CZK
APOSPC5076-25G APOSPC5076-5G APOSPC5076-100G
Methyl-3,5-bis(trifluoromethyl)benzoate 98%
Methyl-3,5-bis(trifluoromethyl)benzoate
Vzorec: C₁₀H₆F₆O₂
MW: 272.15 g/mol
Bod varu: 85…87 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052317
CAS číslo: 26107-80-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR