Tisk…

Methyl difluoroacetate 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC5075-5G PC5075-250G PC5075-50G
APOSPC5075-5GEA 827 CZK
APOSPC5075-5G APOSPC5075-250G APOSPC5075-50G
Methyl difluoroacetate 97%
Methyl difluoroacetate
Vzorec: C₃H₄F₂O₂
MW: 110,06 g/mol
Bod varu: 85…87 °C
Hustota: 1,26 g/cm³ (25 °C)
Bod vzplanutí: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00039276
CAS číslo: 433-53-4
EINECS: 207-089-7
UN: 2924
ADR: 3+8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR