α,α,α-Trifluoro-p-xylene 96+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethyl)toulene, 1-Methyl-4-(trifluoromethyl)benzene, 4-Methylbenzotrifluoride
PC5067C-25G
PC5067C-5G
PC5067C-100G
APOSPC5067C-25GEA
3740
CZK
APOSPC5067C-25G
APOSPC5067C-5G
APOSPC5067C-100G
α,α,α-Trifluoro-p-xylene 96+%
α,α,α-Trifluoro-p-xylene
Vzorec:
C₈H₇F₃ MW: 160.14 Bod varu: 129 °C Hustota: 1.144 g/cm³ (25 °C) Bod vzplanutí: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075476 CAS číslo: 6140-17-6 EINECS: 228-124-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...