Tisk…

2,3,5-Trifluoroiodobenzene

Dodavatel: Apollo Scientific
Varování

PC5066-1G PC5066-5G
APOSPC5066-1GEA 3740 CZK
APOSPC5066-1G APOSPC5066-5G
2,3,5-Trifluoroiodobenzene
2,3,5-Trifluoroiodobenzene
Vzorec: C₆H₂F₃I
Storage Temperature: Refrigerator
MDL Number: MFCD08532463
CAS číslo: 622379-51-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR