Tisk…

(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

PC5063E-100MG PC5063E-1G PC5063E-5G
APOSPC5063E-100MGEA 1040 CZK
APOSPC5063E-100MG APOSPC5063E-1G APOSPC5063E-5G
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₉F₃O₃
MW: 234,17 g/mol
Bod varu: 95…97 °C/0,05mmHg
Bod tání: 46…49 °C
Hustota: 1,303 g/cm³ (25 °C)
Bod vzplanutí: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064200
CAS číslo: 17257-71-5
EINECS: 241-292-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR