Tisk…

4-Fluoro-3-methoxyphenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid

PC5053-1G PC5053-5G PC5053-250MG
APOSPC5053-1GEA 705 CZK
APOSPC5053-1G APOSPC5053-5G APOSPC5053-250MG
4-Fluoro-3-methoxyphenylboronic acid 98%
4-Fluoro-3-methoxyphenylboronic acid
Vzorec: C₇H₈BFO₃
MW: 169.95 g/mol
Bod tání: 189…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056358
CAS číslo: 854778-31-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR