Tisk…

4-Nitro-2-(trifluoromethyl)anisole 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Methoxy-4-nitro-2-(trifluoromethyl)benzene, 2-Methoxy-5-nitrobenzotrifluoride

PC5050-100G PC5050-25G PC5050-500G
APOSPC5050-100GEA 3670 CZK
APOSPC5050-100G APOSPC5050-25G APOSPC5050-500G
4-Nitro-2-(trifluoromethyl)anisole 99%
4-Nitro-2-(trifluoromethyl)anisole
Vzorec: C₈H₆F₃NO₃
MW: 221,14 g/mol
Bod tání: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007328
CAS číslo: 654-76-2
EINECS: 211-507-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR