Tisk…

(5-Methyl-3-trifluoromethyl-pyrazol-1-yl)acetic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl)acetic acid

PC50397-10G PC50397-1G PC50397-5G
APOSPC50397-10GEA 12530 CZK
APOSPC50397-10G APOSPC50397-1G APOSPC50397-5G
(5-Methyl-3-trifluoromethyl-pyrazol-1-yl)acetic acid 95%
(5-Methyl-3-trifluoromethyl-pyrazol-1-yl)acetic acid
Vzorec: C₇H₇F₃N₂O₂
MW: 208,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297315
CAS číslo: 345637-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR