(3-Fluoro-4-hydroxyphenyl)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Fluoro-4-hydroxyphenylacetic acid
PC5039-1G
PC5039-25G
PC5039-5G
APOSPC5039-1GEA
830
CZK
APOSPC5039-1G
APOSPC5039-25G
APOSPC5039-5G
(3-Fluoro-4-hydroxyphenyl)acetic acid
(3-Fluoro-4-hydroxyphenyl)acetic acid
Vzorec:
C₈H₇FO₃ MW: 170,14 g/mol Bod tání: 130…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004348 CAS číslo: 458-09-3 EINECS: 207-275-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...