5-Bromo-2-fluorophenylisothiocyanate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC503018-1G
PC503018-25G
PC503018-5G
PC503018-100G
APOSPC503018-1GEA
622
CZK
APOSPC503018-1G
APOSPC503018-25G
APOSPC503018-5G
APOSPC503018-100G
5-Bromo-2-fluorophenylisothiocyanate
5-Bromo-2-fluorophenylisothiocyanate
Vzorec:
C₇H₃BrFNS MW: 232,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09800737 CAS číslo: 1027513-65-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...