Tisk…

4-Bromo-3-fluorophenyl isothiocyanate

Dodavatel: Apollo Scientific
Nebezpečí

PC503016-1G PC503016-25G PC503016-5G
APOSPC503016-1GEA 1370 CZK
APOSPC503016-1G APOSPC503016-25G APOSPC503016-5G
4-Bromo-3-fluorophenyl isothiocyanate
4-Bromo-3-fluorophenyl isothiocyanate
Vzorec: C₇H₃BrFNS
MW: 232,08 g/mol
Storage Temperature: Ambient
CAS číslo: 362690-56-0
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR