Diphenyliodonium trifluoromethanesulphonate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC5029-1G
PC5029-5G
PC5029-10G
APOSPC5029-1GEA
622
CZK
APOSPC5029-1G
APOSPC5029-5G
APOSPC5029-10G
Diphenyliodonium trifluoromethanesulphonate 97%
Diphenyliodonium trifluoromethanesulphonate
Vzorec:
C₁₃H₁₀F₃IO₃S MW: 430,19 g/mol Bod tání: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191356 CAS číslo: 66003-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...