4-Nitro-3-(trifluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
PC502658-25G
PC502658-5G
APOSPC502658-25GEA
6120
CZK
APOSPC502658-25G
APOSPC502658-5G
4-Nitro-3-(trifluoromethyl)benzonitrile
4-Nitro-3-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃F₃N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD05664270 CAS číslo: 320-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...