Tisk…

Methyl-2-fluoro-5-nitrobenzenecarboxylate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-fluoro-5-nitrobenzenecarboxylate, Methyl 2-fluoro-5-nitrobenzoate

PC502595-25G
APOSPC502595-25GEA 1820 CZK
APOSPC502595-25G
Methyl-2-fluoro-5-nitrobenzenecarboxylate 98%
Methyl-2-fluoro-5-nitrobenzenecarboxylate
Vzorec: C₈H₆FNO₄
MW: 199,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203689
CAS číslo: 2965-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR