Tisk…

Methyl-2-fluoro-4-iodobenzoate

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-fluoro-4-iodobenzoate

PC502581-1G PC502581-5G
APOSPC502581-1GEA 3270 CZK
APOSPC502581-1G APOSPC502581-5G
Methyl-2-fluoro-4-iodobenzoate
Methyl-2-fluoro-4-iodobenzoate
Vzorec: C₈H₆FIO₂
MW: 280,0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846098
CAS číslo: 204257-72-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR