Methyl-2-bromo-6-fluorobenzoate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-bromo-6-fluorobenzoate, 2-Bromo-6-fluorobenzoic acid methyl ester
PC502436-1G
PC502436-5G
PC502436-10G
APOSPC502436-1GEA
622
CZK
APOSPC502436-1G
APOSPC502436-5G
APOSPC502436-10G
Methyl-2-bromo-6-fluorobenzoate 98%
Methyl-2-bromo-6-fluorobenzoate
Vzorec:
C₈H₆BrFO₂ MW: 233.04 Storage Temperature: Ambient |
MDL Number:
MFCD10000922 CAS číslo: 820236-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...