Tisk…

2-Bromo-3-(difluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC502422-10G PC502422-1G PC502422-5G
APOSPC502422-10GEA 22420 CZK
APOSPC502422-10G APOSPC502422-1G APOSPC502422-5G
2-Bromo-3-(difluoromethyl)pyridine 95%
2-Bromo-3-(difluoromethyl)pyridine
Vzorec: C₆H₄BrF₂N
MW: 208.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257230
CAS číslo: 1211515-00-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR