5-(Difluoromethyl)-2-fluoropyridine 97%
Dodavatel: Apollo Scientific
PC502322-10G
PC502322-1G
PC502322-250MG
APOSPC502322-10GEA
22520
CZK
APOSPC502322-10G
APOSPC502322-1G
APOSPC502322-250MG
5-(Difluoromethyl)-2-fluoropyridine 97%
5-(Difluoromethyl)-2-fluoropyridine
Vzorec:
C₆H₄F₃N Storage Temperature: Ambient |
MDL Number:
MFCD22199299 CAS číslo: 1374659-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...