3-(Difluoromethyl)-2-fluoropyridine 95%
Dodavatel: Apollo Scientific
PC502292-10G
PC502292-1G
PC502292-5G
APOSPC502292-10GEA
15970
CZK
APOSPC502292-10G
APOSPC502292-1G
APOSPC502292-5G
3-(Difluoromethyl)-2-fluoropyridine 95%
3-(Difluoromethyl)-2-fluoropyridine
Vzorec:
C₆H₄F₃N Storage Temperature: Ambient |
MDL Number:
MFCD22199298 CAS číslo: 1374659-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...