Tisk…

Pentafluorobenzoyl chloride 97%

Dodavatel: Apollo Scientific

Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

PC50171-100G PC50171-25G PC50171-250G
APOSPC50171-100GEA 3000 CZK
APOSPC50171-100G APOSPC50171-25G APOSPC50171-250G
Pentafluorobenzoyl chloride 97%
Pentafluorobenzoyl chloride

Useful reagent for the derivatisation of primary amines, etc for GC analysis.

Vzorec: C₇ClF₅O
Bod varu: 158…159 °C
Hustota: 1,601 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000657
CAS číslo: 2251-50-5
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR