Tisk…

3'-Bromo-5'-fluoroacetophenone 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(3-Bromo-5-fluorophenyl)ethanone

PC501607-10G PC501607-1G PC501607-5G PC501607-25G
APOSPC501607-10GEA 6680 CZK
APOSPC501607-10G APOSPC501607-1G APOSPC501607-5G APOSPC501607-25G
3'-Bromo-5'-fluoroacetophenone 95%
3'-Bromo-5'-fluoroacetophenone
Vzorec: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD11847746
CAS číslo: 105515-20-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR