Tisk…

4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

PC5016-250MG PC5016-1G PC5016-5G
APOSPC5016-250MGEA 1240 CZK
APOSPC5016-250MG APOSPC5016-1G APOSPC5016-5G
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid
Vzorec: C₈H₉BFNO₃
MW: 196,97 g/mol
Bod tání: 226…228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235038
CAS číslo: 874219-19-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR