Tisk…

3'-Methyl-2,2,2-trifluoroacetophenone 95%

Dodavatel: Apollo Scientific

PC501401-10G PC501401-5G
APOSPC501401-10GEA 6220 CZK
APOSPC501401-10G APOSPC501401-5G
3'-Methyl-2,2,2-trifluoroacetophenone 95%
3'-Methyl-2,2,2-trifluoroacetophenone
Vzorec: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD00461893
CAS číslo: 1736-06-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR