4'-Bromo-3'-fluoroacetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-3-fluoro--acetophenone
PC501320-1G
PC501320-5G
PC501320-10G
APOSPC501320-1GEA
622
CZK
APOSPC501320-1G
APOSPC501320-5G
APOSPC501320-10G
4'-Bromo-3'-fluoroacetophenone
4'-Bromo-3'-fluoroacetophenone
Vzorec:
C₈H₆BrFO MW: 217.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00466241 CAS číslo: 304445-49-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...