Tisk…

3-Bromo-5-fluoro-2-iodobenzotrifluoride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-5-fluoro-2-iodo-3-(trifluoromethyl)benzene

PC501312-10G PC501312-1G PC501312-5G
APOSPC501312-10GEA 14060 CZK
APOSPC501312-10G APOSPC501312-1G APOSPC501312-5G
3-Bromo-5-fluoro-2-iodobenzotrifluoride 98%
3-Bromo-5-fluoro-2-iodobenzotrifluoride
Vzorec: C₇H₂BrF₄I
Storage Temperature: Ambient
MDL Number: MFCD09800680
CAS číslo: 1027511-93-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR