Tisk…

3-(Ethoxycarbonyl)-4-fluorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

PC5013-1G PC5013-5G
APOSPC5013-1GEA 3510 CZK
APOSPC5013-1G APOSPC5013-5G
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid 98%
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid
Vzorec: C₉H₁₀BFO₄
MW: 211,9827 g/mol
Bod tání: 149…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061836
CAS číslo: 874219-36-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR